missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ DS-30 Matrix Standard Kit (Dye Set D)
Description
Includes
Contains 4 oligonucleotides labeled respectively with 6FAM™,HEX™,NED™,and ROX™
The DS-30 Matrix Standard kit contains 4 oligonucleotides respectively labeled with 6-FAM™, HEX™, NED™, and ROX™, pre-pooled in a single tube. This Matrix Standard Set is used to generate the “multicomponent matrix” required when analyzing fluorescent dye-labeled DNA fragments on the Applied Biosystems™ Systems. The Data Collection Software for these instruments uses the multicomponent matrix to automatically analyze the four different colored fluorescent dye-labeled samples in a single capillary.
Order Info
Each
Specifications
Specifications
| Shipping Condition | Wet Ice |
| For Use With (Equipment) | SeqStudio 3500 and 3730xl |
| Quantity | 8 runs |
For Research Use Only. Not for use in diagnostic procedures.