Learn More
BUCHI Drying Flask
Description
- Indents reduce accumulation on the flask walls
- Increased efficiency in powder drying
- Standard Taper Joint: 24/40
Spécifications
Spécifications
| Material | Borosilicate Glass (3.3) |
| Type | Drying Flask |
| For Use With (Application) | Powder Drying |
| For Use With (Equipment) | Rotary Evaporators |
| Shape | Pear |
| Standard Taper Joint Number Center | 24/40 |
| Volume (Metric) | 500 mL |