missing translation for 'onlineSavingsMsg'
Learn More

JUSTRITE Drum Storage Cabinets, 100 gal., Manual Close, with Drum Rollers

Numéro de catalogue. 18666762
Change view
Click to view available options
Color:
Blue
Gray
Red
Yellow
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Color
18666762 Blue
18666760 Yellow
18666761 Red
18666763 Gray
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Numéro de catalogue. 18666762 Fournisseur JUSTRITE Code fournisseur 899162

Pour de laide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

For storage of larger quantities of flammable solvents, the heavy-duty Drum Safety Cabinet is available in many styles and sizes.

Built to the same high standards as the smaller Classic Safety Cabinets. An array of drum accessories is available to ensure safety and convenience when storing and handling drums of hazardous materials.

  • 18-gauge (1 mm) double-wall, welded steel with 38 mm of insulating air space for fire resistance.
  • Half-depth shelf adjusts to accommodate a drum with pump or funnel, or stores spare drum accessories.
  • Available in manual or self-close models.
  • Choice of drum rollers to facilitate the movement of heavy drums or removable drum support to keep drums elevated off cabinet sump.
  • Independently fire tested and approved by FM Approvals.
  • High-performance, self-latching doors close easily and securely for maximum protection under fire conditions.
  • U-Loc™ handle comes with a cylinder lock and key set or accepts an optional padlock.
  • Haz-Alert™ reflective warning labels are highly visible under fire conditions.
  • Patented SpillSlope™ galvanized steel shelves direct spills to back and bottom of a leakproof sump.
  • Designed to meet OSHA and NFPA 30 standards. FM-approved and come with a ten-year warranty

Spécifications

Surface Coating Powder-coat Paint
No. of Shelves 1
Width (English) Interior 56 in.
Weight (English) 601 lb.
Weight (Metric) 272.6 kg
Width (Metric) Exterior 1499 mm
Width (English) Exterior 59 in.
Width (Metric) Interior 1422 mm
Product Type Vertical Drum Storage Cabinet
Height (English) Exterior 65 in.
Material Steel (18 ga), Galvanized Steel
Color Blue
Depth (English) Exterior 59 in.
Afficher plus de résultats Afficher moins de résultats