Learn More
Stoelting™ Drill Bit
Numéro de catalogue. 10001086
Change view
Click to view available options
Size:
0.60 mm
Numéro de catalogue. Size
10001086 0.60 mm
1 options
Numéro de catalogue. 10001086 Fournisseur Stoelting™ Code fournisseur 514552
Il en reste null
Stoelting™ Carbide Drill Bits cut cleaner and retain sharp edges longer.
  • Generally used in Stereotaxic Surgery and procedures
  • Suitable for use with the 58650 Flexible Shaft Drill, and the 51449 Micromotor Drill
  • Spherical heads, variety of sizes
  • Packages of 6 bits (all the same size)
Spécifications
For Use With (Application) Neuroscience
Size 0.60 mm
Material Carbide