missing translation for 'onlineSavingsMsg'
Learn More

Drierite™, with indicator, 8 mesh

Catalog No. AC219090050
Change view
Click to view available options
Quantity:
500 g
2 kg
5 kg
Packaging:
Glass Bottle
Plastic Drum
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC219090050 5 kg Plastic Drum
AC219095000 500 g Glass Bottle
AC219090020 2 kg Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC219090050 Supplier Thermo Scientific Chemicals Supplier No. 219090050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Drierite, CaO4S, CAS Number-7778-18-9, 7646-79-9, 5kg, 39.09%, 0.68%, CHEBI:31346, 3.25%, Blue, 01,107, 136.13, GHS Signal Word: Danger | CAS: 7778-18-9 | CaO4S | 136.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Adsorbent, Indicator

Chemical Identifiers

CAS 7778-18-9 , 7646-79-9
Molecular Formula CaO4S
Molecular Weight (g/mol) 136.13
MDL Number MFCD00010912
InChI Key OSGAYBCDTDRGGQ-UHFFFAOYSA-L
PubChem CID 24497
ChEBI CHEBI:31346
SMILES [Ca++].[O-]S([O-])(=O)=O

Specifications

Melting Point 2642°F
Color Blue
Physical Form Granular
Quantity 5 kg
Fieser 01,107
Merck Index 15, 1708
Packaging Plastic Drum
Solubility Information SiO2 and insolubles: 0.58%
R2O 0.29%
IUPAC Name calcium;sulfate
Formula Weight 136.13
Chemical Name or Material Drierite™, With indicator, 8 Mesh
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
May cause cancer by inhalation.
Suspected of causing genetic defects.
May damage fertility.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.
Very toxic to aquatic life with long lasting effect
Precautionary Statement GHS P Statement:
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid release to the environment.
Obtain special instructions before use.
Use personal protective equipment as required.
Do not handle until all safety precautions have been read and understood.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only