missing translation for 'onlineSavingsMsg'
Learn More

Drierite™, indicating (CaSO{4})

Catalog No. AA43065P2
Change view
Click to view available options
Quantity:
1 lb.
5 lb.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA43065P2 1 lb.
AA43065P5 5 lb.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA43065P2 Supplier Thermo Scientific Chemicals Supplier No. 043065P2

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7778-18-9 | CaO4S | 136.13 g/mol

Drierite™, indicating (CaSO{4}) is used as a desiccant. It is used as a drying agent utilized for the efficient and rapid drying of air, industrial gases, refrigerants, organic liquids and solids. Further, it is used to maintain a dry atmosphere in storage spaces, vaults, commercial packages, and other enclosures. In addition to this, it is involved in the protection of hygroscopic materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7778-18-9
Molecular Formula CaO4S
Molecular Weight (g/mol) 136.13
MDL Number MFCD00010912
InChI Key OSGAYBCDTDRGGQ-UHFFFAOYSA-L
PubChem CID 24497
ChEBI CHEBI:31346
IUPAC Name calcium sulfate
SMILES [Ca++].[O-]S([O-])(=O)=O

Specifications

Color Blue
Physical Form Granular
Quantity 1 lb.
Odor Odorless
Solubility Information Soluble in water.
Sensitivity Hygroscopic
Grade Laboratory
Chemical Name or Material Drierite
Hazard Category H317-H334-H341-H350i-H360F
Precautionary Statement P201-P202-P261-P272-P280g-P281-P285-P302+P352-P304+P341-P308+P313-P333+P313-P342+P311-P363-P501c
DOTInformation Hazard Class: 9; Packaging Group: III
EINECSNumber 231-900-3
RTECSNumber WS6920000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only