missing translation for 'onlineSavingsMsg'
Learn More

Dried Ferrous Sulfate, Powder, USP, 86-89%, Spectrum™ Chemical

Catalog No. 18602353
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18602353 500 g Poly Bottle
18602354 2.5 kg Poly Pail
18602355 12 kg Poly Pail
18602356 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18602353 Supplier Spectrum Chemical Mfg Cor Supplier No. F1065500GM
Only null left

F1065, 13463-43-9, FeSO4·xH2O

Spectrum™ Chemical Dried Ferrous Sulfate, Powder, USP is used to prevent or treat low levels of iron in the blood. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 13463-43-9
Molecular Formula FeH2O5S
Molecular Weight (g/mol) 169.92
InChI Key XBDUTCVQJHJTQZ-UHFFFAOYSA-L
IUPAC Name λ²-iron(2+) hydrate sulfate
SMILES O.[Fe++].[O-]S([O-])(=O)=O

Specifications

CAS Max % 100%
Quantity 500 g
Percent Purity 86 to 89%
Grade USP
Packaging Poly Bottle