missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ DreamTaq PCR Master Mixes (2X)

Catalog No. FERK1082
Change view
Click to view available options
Color:
Colorless
Green
Quantity:
4 x 1.25 mL
20 x 1.25 mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Quantity
FERK1082 Green 20 x 1.25 mL
FERK1071 Colorless 4 x 1.25 mL
FERK1072 Colorless 20 x 1.25 mL
FERK1081 Green 4 x 1.25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. FERK1082 Supplier Thermo Scientific™ Supplier No. K1082

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Thermo Scientific DreamTaq PCR Master Mix (2X) is a ready-to-use solution containing DreamTaq DNA Polymerase, optimized DreamTaq buffer, MgCl2, and dNTPs.

Thermo Scientific DreamTaq PCR Master Mix (2X), available in colorless and green formats, is a ready-to-use solution containing DreamTaq DNA Polymerase, optimized DreamTaq buffer, MgCl2, and dNTPs. This pre-mixed formulation saves time and reduces contamination due to the fewer pipetting steps required for PCR setup.

The master mix retains all features of DreamTaq DNA Polymerase and is capable of robust amplification up to 6 kb from genomic DNA and up to 20 kb from viral DNA. It ensures higher sensitivity, longer PCR products, and higher yields compared to conventional Taq DNA polymerase. Extensive optimization of reaction conditions is not required.

DreamTaq Green PCR Master Mix (2X) additionally includes 2X DreamTaq Green Buffer with a density reagent and two dyes for monitoring electrophoresis progress.

Features

  • Robust amplification with minimal optimization
  • High yields of PCR products
  • Higher sensitivity compared to conventional Taq DNA polymerase
  • Amplification of targets up to 6 kb from genomic DNA and up to 20 kb from viral DNA
  • Incorporates modified nucleotides, but does not incorporate dUTP
  • Direct loading of PCR product on gel with green format

Applications:

  • Routine genomic and plasmid DNA amplification
  • Genotyping
  • Colony PCR and clone screening
  • High-throughput PCR screening
  • TA cloning workflows

Specifications

Concentration 2X
Content And Storage • 2X DreamTaq Green PCR Master Mix, 20 x 1.25 mL
• Nuclease-free water, 20 x 1.25 mL

Store at -20°C.
GC-Rich PCR Performance Low
Polymerase DreamTaq DNA Polymerase
Reaction Speed Standard
Quantity 20 x 1.25 mL
Shipping Condition Dry Ice
For Use With (Application) Standard PCR
Fidelity (vs. Taq) 1X
Hot Start No
No. of Reactions 1000 Reactions
Overhang 3'-A
Reaction Format SuperMix or Master Mix
Color Green
Show More Show Less
What is the error rate of DreamTaq DNA Polymerases?

The error rate of DreamTaq DNA Polymerase is the same as that of Taq DNA Polymerase: 2.2x10^-5 errors per nt per cycle.

For Research Use Only. Not for use in diagnostic procedures.