missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ DreamTaq™ Hot Start DNA Polymerase

Catalog No. FEREP1714
Change view
Click to view available options
Color:
Colorless
Green
Quantity:
200 U
2500 U
4 x 2500 U
500 U
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Quantity
FEREP1714 Green 4 x 2500 U
FEREP1701 Colorless 200 U
FEREP1702 Colorless 500 U
FEREP1704 Colorless 4 x 2500 U
FEREP1711 Green 200 U
FEREP1712 Green 500 U
FEREP1713 Green 2500 U
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. FEREP1714 Supplier Thermo Scientific™ Supplier No. EP1714

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Thermo Scientific DreamTaq Hot Start DNA Polymerase is an enhanced hot start Taq DNA polymerase that enables higher PCR specificity, sensitivity, and yields compared to conventional hot start Taq DNA polymerases.

Thermo Scientific DreamTaq Hot Start DNA Polymerase, available in colorless and green formats, is an enhanced hot start Taq DNA polymerase that enables higher PCR specificity, sensitivity, and yields compared to conventional hot start Taq DNA polymerases.

DreamTaq Hot Start DNA Polymerase employs antibody-based inhibition of DNA polymerase activity at ambient temperatures to prevent the amplification of non-specific products prior to the amplification step. With DreamTaq Hot Start DNA Polymerase, reactions can be set up at room temperature using the same protocol and cycling conditions as conventional Taq DNA polymerases. The enzyme is supplied with an optimized 10X DreamTaq Buffer containing magnesium chloride, which eliminates the need for extensive optimization of reaction conditions. DreamTaq Hot Start DNA Polymerase is also available with pre-added density reagent and electrophoresis tracking dyes, as well as in master mix format.

The 10X DreamTaq Green Buffer also includes a density reagent and two tracking dyes for convenient direct loading of PCR products on a gel.

DreamTaq Hot Start DNA Polymerase is formulated to enhance productivity through:

  • Better reaction outcomes
    • Higher yield of target amplicons from low template amounts
    • Increased reaction specificity due to reliable hot-start technology
    • Wider range of amplicon lengths with routine amplification of genomic DNA fragments up to 6 kb
  • Enhanced convenience
    • Room temperature reaction setup
    • Minimized optimization of Mg2+ concentration and of primer annealing temperatures due to optimized reaction buffer
    • Direct loading of PCR products on a gel with DreamTaq Green Buffer

Applications:

  • Routine genomic and plasmid DNA amplification
  • Genotyping
  • Colony PCR and clone screening
  • High-throughput PCR screening
  • TA cloning workflows

Recommended Storage

Store at -20C.

Specifications

Concentration 5 U/μL
Content And Storage • DreamTaq Hot Start DNA Polymerase (5 U/μL), 20 x 100 μL
• 10X DreamTaq Green Buffer (with 20 mM MgCl2), 40 x 1.25 mL

Store at -20°C.
GC-Rich PCR Performance Low
Polymerase DreamTaq DNA Polymerase
Reaction Speed Standard
Quantity 4 x 2500 U
Shipping Condition Dry Ice
For Use With (Application) Hot-start PCR
Hot Start Built-In Hot Start
Overhang 3'-A
Reaction Format Separate Components
Color Green
Show More Show Less
Which nucleotide analogues can be used with DreamTaq Hot Start DNA Polymerase?

DreamTaq Hot Start DNA Polymerase can incorporate dUTP and a variety of modified nucleotides, such as: dITP, 7-deaza-dGTP, fluorescein-12-dUTP, Biotin-11-dUTP, dm5CTP, alpha-thio-dCTP, and aminoallyl-dUTP.

Are DreamTaq buffers the same for DreamTaq DNA Polymerase and DreamTaq Hot Start DNA Polymerase?

Yes. 10X DreamTaq Buffer and 10X DreamTaq Green Buffer are the same for both polymerases.

My experiments require extra DreamTaq Buffer. Do you offer the buffer as a separate product?

Yes, DreamTaq Buffer (Cat. No. B65, 4 x 1.25 mL) and DreamTaq Green Buffer (Cat. No. B71, 4 x 1.25 mL) are available as stand-alone products.

Can DreamTaq Hot Start DNA Polymerase be used for amplification of bisulfite converted DNA?

Yes. DreamTaq Hot Start DNA Polymerase can read uracil in the template strand and therefore can be used for amplification of bisulfite converted DNA.

What is the limit of amplicon size using DreamTaq Hot Start DNA Polymerase?

DreamTaq Hot Start DNA Polymerase amplifies up to 6 kb from human genomic DNA and up to 20 kb from lambda DNA with high yields and specificity. Amplification of even longer fragments up to 9 kb from human genomic DNA has been demonstrated, but may require additional optimization of reaction conditions and primer design.

Does DreamTaq Hot Start DNA Polymerase add the non-template dependent 3'-A overhang?

Yes. DreamTaq Hot Start DNA Polymerase generates 3'-A overhangs.