Learn More
Dräger™ Short-Term Detector Tubes - Hydrocarbons
Catalog No. 17985239
Change view
Click to view available options
Components Measured:
Hydrocarbon 2/a
Measurement Range:
2 to 24mg/L
Catalog No. Components Measured Measurement Range
17985239 Hydrocarbon 2/a 2 to 24mg/L
1 options
Catalog No. 17985239 Supplier Dräger™ Supplier No. 8103581
  1. Extension of measuring range listed is possible for some detector tubes. Call Fisher Scientific™ at 1-800-772-6733 for more information.
  2. Safety Equipment Institute (SEI) certified.
Specifications
Components Measured Hydrocarbon 2/a
Humidity Range 1 to 25mg/L
Measurement Range 2 to 24mg/L
No. of Strokes 3
Quantity 10
Response Time 5 min.
Temperature Range (English) 32°F to 104°F
Temperature Range (Metric) 0°C to 40°C