missing translation for 'onlineSavingsMsg'
Learn More

Dräger™ Short-Term Detector Tubes - De Tube Hydrogen

Catalog No. 17985244
Change view
Click to view available options
Components Measured:
Hydrogen Fluoride 1.5/b (gas or vapor)
Hydrogen Peroxide 0.1/a (gas or vapor)
Measurement Range:
0.1 to 3ppm
15ppm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Components Measured Measurement Range
17985244 Hydrogen Peroxide 0.1/a (gas or vapor) 0.1 to 3ppm
17985243 Hydrogen Fluoride 1.5/b (gas or vapor) 15ppm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 17985244 Supplier Dräger™ Supplier No. 8101041
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For making spot measurments for various gases and vapors

  • Glass vial
  • Calibrated 100mL sample of air drawn through tube with Drager Accuro™ bellows pump

Order Info

Extension of measuring range listed is possible for some detector tubes. Call Fisher Scientific™ at 1-800-772-6733 for more information.

Specifications

Components Measured Hydrogen Peroxide 0.1/a (gas or vapor)
For Use With (Equipment) Accuro Drager Gas Detector Pump
Type Gas Detection Tube
Measurement Range 0.1 to 3ppm
Quantity 10/Box