Learn More
Dräger™ Pac™ III Sensors
Provide fast and accurate responses to hazardous gas levels
Supplier: Dräger™ 6808582
Description
- For use with Pac™ III single gas monitors
Identifiants chimiques
| 700 to 1300mbar |
Spécifications
| Phosgene (COCL2) | |
| 0 to 3 | |
| 0.01 | |
| -20°C to +50°C |
| 10 to 95% | |
| 700 to 1300mbar | |
| -4°F to +122°F | |
| 6 Months |