missing translation for 'onlineSavingsMsg'
Learn More

Dräger™ CMS Chips: Vinyl Chloride

Suitable for 10 measurements of one gas

$225.45

Spécifications

Air Pressure 700 to 1100 hpa
Type Vinyl Chloride CMS Chip
Humidity Range 1 to 30mg/L
Temperature Range (English) 32°F to 104°F
Temperature Range (Metric) 0°C to 40°C
voir les spécifications

Produits 1
Numéro de catalogue Numéro du manufacturier. Measurement Range Response Time Prix Quantity & Availability  
Numéro de catalogue Numéro du manufacturier. Measurement Range Response Time Prix Quantity & Availability  
19034578
Afficher les documents Détails de la promotion
Dräger™
6406230
10 to 250 45 to 100 sec.
paquet for $225.45
Il en reste null
 
Description

Description

Automatically supplies the analyzer with the calibration data and measurement range of the respective gas.
Spécifications

Spécifications

700 to 1100 hpa
1 to 30mg/L
0°C to 40°C
Vinyl Chloride CMS Chip
32°F to 104°F
Videos
FDS
Documents

Documents