Learn More
Dräger™ CMS Chips: Acetic Acid
Catalog No. 19040947
Change view
Click to view available options
Measurement Range:
2 to 50
Catalog No. Measurement Range
19040947 2 to 50
1 options
Catalog No. 19040947 Supplier Dräger™ Supplier No. 6406330
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Suitable for 10 measurements of one gas
Automatically supplies the analyzer with the calibration data and measurement range of the respective gas.
Specifications
Air Pressure 700 to 1100 hpa
Type Acetic Acid CMS Chip
Humidity Range 1 to 30mg/L
Measurement Range 2 to 50
Response Time 40 to 300 sec.
Temperature Range (English) 32°F to 104°F
Temperature Range (Metric) 0°C to 40°C