missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ DPTN
Description
DPTN is a potent and selective A3 adenosine receptor antagonist (Ki values are in the range 0.36 - 1.65 nM (hA3AR) and 1.6 - 8.53 nM (rA3AR)). Reduces intrapulmonary pressure when combined with Dexamethasone (Cat. No. 1126).
Spécifications
Spécifications
| Chemical Name or Material | N-[4-(3,5-Dimethylphenyl)-5-(4-pyridinyl)-2-thiazolyl]-3-pyridinecarboxamide dihydrochloride |
| CAS | 325767-87-1 |
| Quantity | 50 mg |
| Target | Adenosine A3 R |
| Molecular Formula | C22H18N4OS.2HCl |