missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DPCD Monoclonal Antibody (K1E042_4B5), Invitrogen™
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA556055
Description
Sequence of this protein is as follows: RTAQKTALLQ DGRRKVHYLF PDGKEMAEEY DEKTSELLVR KWRVKSALGA MGQWQLEVGD PAPLGAGNLG PELIKESNAN PIFMRKDTKM SFQWRIRNLP YPKDVYSVSV DQKERCIIVR TTNKKYYKKF SIPDLDRHQL PLDDALLSFA HANCTLIISY QKPKEVVVAE SELQKELKKV KTAHSNDGDC
May play a role in the formation or function of ciliated cells.Identifiants chimiques
| DPCD | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| 5330431N19Rik; deleted in a mouse model of primary ciliary dyskinesia; deleted in primary ciliary dyskinesia; deleted in primary ciliary dyskinesia homolog (mouse); DPCD; Ndac; neighbor of Dac; protein DPCD; RGD1307648; RP11-529I10.4; RP11-529I104 | |
| Recombinant human DPCD (immunogen range is 11-200). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| DPCD |
| K1E042_4B5 | |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Mouse | |
| Human, Mouse | |
| Q8BPA8, Q9BVM2 | |
| 226162, 25911 | |
| IgG2b | |
| Protein A | |
| Antibody |
Spécifications
| DPCD | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| Q8BPA8, Q9BVM2 | |
| DPCD | |
| Recombinant human DPCD (immunogen range is 11-200). | |
| 200 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG2b |
| Immunoprecipitation, Western Blot | |
| K1E042_4B5 | |
| Unconjugated | |
| DPCD | |
| 5330431N19Rik; deleted in a mouse model of primary ciliary dyskinesia; deleted in primary ciliary dyskinesia; deleted in primary ciliary dyskinesia homolog (mouse); DPCD; Ndac; neighbor of Dac; protein DPCD; RGD1307648; RP11-529I10.4; RP11-529I104 | |
| Mouse | |
| Protein A | |
| RUO | |
| 226162, 25911 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |