missing translation for 'onlineSavingsMsg'
Learn More

Doxorubicin hydrochloride

Catalog No. AAJ64000MF
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ64000MF 50 mg
AAJ64000MA 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ64000MF Supplier Thermo Scientific Chemicals Supplier No. J64000MF

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Doxorubicin hydrochloride, CAS # 25316-40-9, is the hydrochloride salt of doxorubicin, isolated from the bacterium Streptomyces peucetius, and is considered to be an anthracycline antibiotic with antineoplastic activity. | CAS: 25316-40-9 | C27H30ClNO11 | 579.98 g/mol

Antitumor antibiotic agent that inhibits DNA topoisomerase II.Doxorubicin hydrochloride is used as an antineoplastic, antibiotic agent that inhibits DNA topoisomerase. It also inhibits nucleic acid synthesis and induces apoptosis. It posses fluorescent property, which enable it for the measurement of drug efflux pump activities and the role of subcellular organelles (golgi and lysosome) in the sequestration of drugs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Doxorubicin hydrochloride, isolated from the bacterium Streptomyces peucetius, is an anthracycline antibiotic with antineoplastic activity and is related to daunorubicin
  • It can intercalate between base pairs in the DNA helix preventing DNA replication
  • It also inhibits the nucleic acid synthesis and can induce apoptosis
  • It can inhibit DNA topoisomerase II, resulting in an increased and stabilized cleavable enzyme-DNA linked complex during DNA replication, and prevents nucleotide strand ligation after double-strand breakage
  • Doxorubicin hydrochloride forms oxygen free radicals that result in cytotoxicity and lead to the lipid peroxidation of cell membrane lipids

Application

  • This compound displays a fluorescent property allowing the measurement of drug efflux pump activity and examining the role of subcellular organelles (Golgi complex and lysosome) in the sequestration of drugs
  • It has been used in cell viability assays
  • It is used as a drug in polymeric PLGA-based microparticulate drug delivery and to develop in vitro doxorubicin-resistant HepG2 cells (HepG2-DR) and K562 cells (K562-DR)
  • It is a substrate of the MRP1 transporter

Chemical Identifiers

CAS 25316-40-9
Molecular Formula C27H30ClNO11
Molecular Weight (g/mol) 579.98
MDL Number MFCD00077757,MFCD00077757,MFCD00941448
InChI Key MWWSFMDVAYGXBV-FGBJBKNOSA-N
Synonym doxorubicin hydrochloride, adriamycin, doxorubicin hcl, adriacin, adriblastina, rubex, adriamycin hydrochloride, adriblastin, adriamycin pfs
PubChem CID 129626538
IUPAC Name hydrogen (8S,10S)-10-[(4-amino-5-hydroxy-6-methyloxan-2-yl)oxy]-6,8,11-trihydroxy-8-(2-hydroxyacetyl)-1-methoxy-5,7,8,9,10,12-hexahydrotetracene-5,12-dione chloride
SMILES [H+].[Cl-].COC1=CC=CC2=C1C(=O)C1=C(O)C3=C(C[C@](O)(C[C@@H]3OC3CC(N)C(O)C(C)O3)C(=O)CO)C(O)=C1C2=O

Specifications

Melting Point 216°C (decomposition)
Color Orange-Red
Quantity 50 mg
Beilstein 4229251
Merck Index 14,3439
Solubility Information Soluble in water.
Formula Weight 579.99
Physical Form Solid
Chemical Name or Material Doxorubicin hydrochloride
Hazard Category H302-H315-H318-H335-H340-H350-H361fd
Hazard Statement GHS H Statement
H350-H302
May cause cancer.
Harmful if swallowed.
Precautionary Statement P201-P202-P261-P264b-P270-P271-P281-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P310-P330-P332+P313-P362-P501c
EINECSNumber 246-818-3
RTECSNumber QI9295900
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only