missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ Dowex™ 1X8 Chloride Form, 50-100 mesh

Catalog No. 111015206
Change view
Click to view available options
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
111015206 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015206 Supplier MilliporeSigma™ Supelco™ Supplier No. 217417100G
Only null left

This ion-exchange resin has not been specially processed nor cleaned. It is suggested that it be treated to a suitable preliminary elution and wash.

Dowex™ 1X8 is a strongly basic anion exchange resin which is used as an absorbent in solid phase extraction.

Specifications

Particle Size 50/100 Mesh
Packaging Poly Bottle
pH 0 to 14
Quantity 100 g
Product Line DOWEX
Capacity (Metric) 1.2 meq/mL by wetted bed volume
MDL Number MFCD00132718
CAS 69011-19-4

DOWEX is a registered trademark of The Dow Chemical Company or an affiliated company of Dow