missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ Dowex™ 1X2 Chloride Form, 200-400 mesh

Catalog No. 111015195
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015195 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015195 Supplier MilliporeSigma™ Supelco™ Supplier No. 217395500G

This ion-exchange resin has not been specially processed nor cleaned. It is suggested that it be treated to a suitable preliminary elution and wash.

Dowex™ 1X2 is an anion exchange resin.

Specifications

Particle Size 200/400 Mesh
Packaging Poly Bottle
pH 0 to 14
Quantity 500 g
Product Line DOWEX
Capacity (Metric) 65 to 75 % water retention capacity
MDL Number MFCD00146763
CAS 69011-19-4

DOWEX is a registered trademark of The Dow Chemical Company or an affiliated company of Dow