missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Supelco™ Dowex™ 1X2 Chloride Form, 100-200 mesh
Description
Dowex™ 1X2 is an anion exchange resin.
Specifications
Specifications
| Particle Size | 100/200 Mesh |
| Packaging | Poly Bottle |
| pH | 0 to 14 |
| Quantity | 100 g |
| Product Line | DOWEX |
| Capacity (Metric) | 0.7 meq/mL by wetted bed volume |
| MDL Number | MFCD00146763 |
| CAS | 69011-19-4 |
DOWEX is a registered trademark of The Dow Chemical Company or an affiliated company of Dow