missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Dopamine Transporter Polyclonal Antibody
GREENER_CHOICE

Numéro de catalogue. PIPA578382
Modifier l'affichage
Click to view available options
Quantité:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité
PIPA578382 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. PIPA578382 Fournisseur Invitrogen™ Code fournisseur PA578382
Saison des économies
Afficher toutes les promotions

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Rabbit Polyclonal Antibody

Positive Control: rat olfactory bulb, mouse olfactory bulb Predicted Reactivity: Human (93%), Dog (86%), Rhesus Monkey (93%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The Dopamine Transporter (DAT) is responsible for the reaccumulation of dopamine after it has been released. DAT antibodies and antibodies for other markers of catecholamine biosynthesis are widely used as markers for dopaminergic and noradrenergic neurons in a variety of applications including depression, schizophrenia, Parkinson's disease and drug abuse.

Spécifications

Antigène Dopamine Transporter
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.35 mg/mL
Conjugué Unconjugated
Formule PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Gène Slc6a3
Numéro d’acquisition du gène P23977, Q01959, Q61327
Alias du gène DA transporter; Dat; Dat1; dopamine transporter; dopamine transporter 1; PKDYS; SLC6A3; Sodium-dependent dopamine transporter; solute carrier family 6 (neurotransmitter transporter), member 3; solute carrier family 6 (neurotransmitter transporter, dopamine), member 3; solute carrier family 6 member 3; solute carrier family 6, member 3
Symboles de gène Slc6a3
Espèces hôtes Rabbit
Immunogène Recombinant protein encompassing a sequence within the C-terminus region of Mouse Dopamine Transporter. The exact sequence is proprietary.
Méthode de purification Antigen affinity chromatography
Quantité 100 μL
Statut réglementaire RUO
Primaire ou secondaire Primary
ID du gène (Entrez) 13162, 24898, 6531, 80787
Espèces cibles Human, Mouse, Rat, Zebrafish
Contenu et stockage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Type de produit Antibody
Forme Liquid
Isotype IgG
Afficher plus de résultats Afficher moins de résultats