missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Donkey anti-Rat IgG (H+L) Highly Cross-Adsorbed Secondary Antibody, Alexa Fluor™ 647

Numéro de catalogue. A78947
Encompass
Modifier l'affichage
Click to view available options
Quantité:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité
A78947 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. A78947 Fournisseur Invitrogen™ Code fournisseur A78947

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Donkey Polyclonal Secondary Antibody

This donkey anti-rat IgG (H+L) whole secondary antibody has been affinity-purified and shows minimal cross-reactivity to bovine, chicken, goat, guinea pig, hamster, horse, human, mouse, rabbit, and sheep serum proteins. Cross-adsorption or pre-adsorption is a purification step to increase specificity of the antibody resulting in higher sensitivity and less background staining. The secondary antibody solution is passed through a column matrix containing immobilized serum proteins from potentially cross-reactive species. Only the nonspecific-binding secondary antibodies are captured in the column, and the highly specific secondaries flow through. The benefits of this extra step are apparent in multiplexing/multicolor-staining experiments (e.g. flow cytometry) where there is potential cross-reactivity with other primary antibodies or in tissue/cell fluorescent staining experiments where there may be the presence of endogenous immunoglobulins. Alexa Fluor dyes are among the most trusted fluorescent dyes available today. Invitrogen™ Alexa Fluor 647 dye is a near-infrared-fluorescent dye with excitation ideally suited to the 647 nm laser line. For stable signal generation in imaging and flow cytometry, Alexa Fluor 647 dye is pH-insensitive over a wide molar range. Probes with high fluorescence quantum yield and high photostability allow detection of low-abundance biological structures with great sensitivity. Alexa Fluor 647 dye molecules can be attached to proteins at high...

Anti-Rat secondary antibodies are affinity-purified antibodies with well-characterized specificity for rat immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.

Spécifications

Antigène Rat IgG (H+L) Highly Cross-Adsorbed
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 2 mg/mL
Conjugué Alexa Fluor 647
Formule PBS with 5mM sodium azide; pH 7.5
Espèces hôtes Donkey
Immunogène Gamma Immunoglobins Heavy and Light chains.
Méthode de purification Purified
Quantité 1 mg
Statut réglementaire RUO
Primaire ou secondaire Secondary
Espèces cibles Rat
Contenu et stockage 4°C, store in dark, DO NOT FREEZE!
Type de produit Secondary Antibody
Forme Liquid
Isotype IgG
Afficher plus de résultats Afficher moins de résultats