missing translation for 'onlineSavingsMsg'
Learn More

IgG Donkey anti-Rat, Biotin, Polyclonal, Secondary Antibody, MilliporeSigma™

Numéro de catalogue. AP189BMI
Modifier l'affichage
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
AP189BMI 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. AP189BMI Fournisseur MilliporeSigma Code fournisseur AP189B

Donkey Polyclonal Antibody

Detect Donkey Rat IgG using this Donkey anti-Rat IgG Antibody, biotin conjugate, Species Adsorbed validated for use in ELISA and western blotting.

Spécifications

Antigen Donkey anti-Rat IgG Species adsorbed
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Biotin
Host Species Donkey
Quantity 500 μL
Primary or Secondary Secondary
Target Species Rat

For Research Use Only