missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Donkey anti-Rabbit IgG (H+L) Cross-Adsorbed Secondary Antibody, DyLight™ 680

Catalog No. PISA510042
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PISA510042 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PISA510042 Supplier Invitrogen™ Supplier No. SA510042

Donkey Polyclonal Secondary Antibody

This antibody is cross-adsorbed and exhibits minimum reactivity to bovine, chicken, goat, human, mouse, pig and rat. For Western blot applications, 5% non-fat dry milk in PBST or TBST is recommended for blocking and incubation of antibodies. BSA is not recommended.

Anti-Rabbit secondary antibodies are affinity-purified antibodies with well-characterized specificity for rabbit immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.

Specifications

Antigen Rabbit IgG (H+L) Cross-Adsorbed
Applications Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate DyLight 680
Formulation PBS with 0.2% BSA and 0.09% sodium azide; pH 6.8 to 7.4
Host Species Donkey
Immunogen Rabbit IgG-heavy and light chain.
Purification Method Antigen affinity chromatography
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Rabbit
Content And Storage 4°C
Product Type Secondary Antibody
Form Liquid
Isotype IgG
Show More Show Less