missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Donkey anti-Mouse IgG (H+L) Secondary Antibody, Biotin

Numéro de catalogue. A16015
Encompass
Modifier l'affichage
Click to view available options
Quantité:
2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité
A16015 2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. A16015 Fournisseur Invitrogen™ Code fournisseur A16015

Donkey Polyclonal Secondary Antibody

The sensitivity of each lot of antibody is confirmed using ELISA. The specificity of each lot of antibody is confirmed by immunoelectrophoresis (IEP).

Anti-Mouse secondary antibodies are affinity-purified antibodies with well-characterized specificity for mouse immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.

Spécifications

Antigène Mouse IgG (H+L)
Applications ELISA, Flow Cytometry, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.5 mg/mL
Conjugué Biotin
Formule PBS with 1% BSA and 0.05% sodium azide; pH 7.2
Espèces hôtes Donkey
Immunogène Gamma Immunoglobins Heavy and Light chains.
Méthode de purification Affinity chromatography
Quantité 2 mg
Statut réglementaire RUO
Primaire ou secondaire Secondary
Espèces cibles Mouse
Contenu et stockage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Type de produit Secondary Antibody
Forme Liquid
Isotype IgG
Afficher plus de résultats Afficher moins de résultats