missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Donkey anti-Chicken IgY (H+L) Highly Cross Adsorbed Secondary Antibody, Alexa Fluor™ 488

Catalog No. A78948
Encompass
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A78948 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A78948 Supplier Invitrogen™ Supplier No. A78948

Donkey Polyclonal Secondary Antibody

This donkey anti-chicken IgY (H+L) whole secondary antibody has been affinity-purified and shows minimum cross-reactivity to bovine, goat, guinea pig, syrian hamster, horse, human, mouse, rabbit, rat and sheep serum proteins. Cross-adsorption or pre-adsorption is a purification step to increase specificity of the antibody resulting in higher sensitivity and less background staining. The secondary antibody solution is passed through a column matrix containing immobilized serum proteins from potentially cross-reactive species. Only the nonspecific-binding secondary antibodies are captured in the column, and the highly specific secondary antibodies flow through. The benefits of this extra step are apparent in multiplexing/multicolor-staining experiments (e.g. flow cytometry) where there is potential cross-reactivity with other primary antibodies or in tissue/cell fluorescent staining experiments where there may be the presence of endogenous immunoglobulins. Alexa Fluor dyes are among the most trusted fluorescent dyes available today. Invitrogen™ Alexa Fluor 488 dye is a bright, green-fluorescent dye with excitation ideally suited to the 488 nm laser line. For stable signal generation in imaging and flow cytometry, Alexa Fluor 488 dye is pH-insensitive over a wide molar range. Probes with high fluorescence quantum yield and high photostability allow detection of low-abundance biological structures with great sensitivity. Alexa Fluor 488 dye molecules can be attached to p...

Anti-Chicken secondary antibodies are affinity-purified antibodies with well-characterized specificity for the chicken immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.

Specifications

Antigen Chicken IgY (H+L) Highly Cross Adsorbed
Applications Peptide Array, Flow Cytometry, Immunohistochemistry, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 2 mg/mL
Conjugate Alexa Fluor 488
Formulation PBS with 5mM sodium azide; pH 7.5
Host Species Donkey
Immunogen Purified Chicken IgY, Heavy and Light chains.
Purification Method Purified
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Chicken
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Secondary Antibody
Form Liquid
Isotype IgG
Show More Show Less