missing translation for 'onlineSavingsMsg'
Learn More

Domperidone 98.0+%, TCI America™

Catalog No. D4125200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D4125200MG 200 mg
D41251G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D4125200MG Supplier TCI America Supplier No. D4125200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 57808-66-9
Molecular Formula C22H24ClN5O2
Molecular Weight (g/mol) 425.917
MDL Number MFCD00069256
InChI Key FGXWKSZFVQUSTL-UHFFFAOYSA-N
Synonym domperidone, motilium, nauzelin, domperidonum, domperidona, domperidon, domperidonum inn-latin, domperidona inn-spanish, motillium, motinorm
PubChem CID 3151
ChEBI CHEBI:31515
IUPAC Name 6-chloro-3-[1-[3-(2-oxo-3H-benzimidazol-1-yl)propyl]piperidin-4-yl]-1H-benzimidazol-2-one
SMILES C1CN(CCC1N2C3=C(C=C(C=C3)Cl)NC2=O)CCCN4C5=CC=CC=C5NC4=O

Specifications

Melting Point 243°C
Color White-Yellow
Quantity 200 mg
Formula Weight 425.92
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Domperidone
RTECSNumber DE2275900
TSCA No