missing translation for 'onlineSavingsMsg'
Learn More

Domiphen bromide, 97%

Catalog No. AAL1293022
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1293022 100 g
AAL1293014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1293022 Supplier Thermo Scientific Chemicals Supplier No. L1293022

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 538-71-6 | C22H40BrNO | 414.472 g/mol

Domiphen bromide is employed as a phase transfer catalyst in synthetic chemistry. It is used as a quaternary ammonium cation with cationic surfactant properties. Further, it is used as an antiseptic and an active pharmaceutical ingredient.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Domiphen bromide is employed as a phase transfer catalyst in synthetic chemistry. It is used as a quaternary ammonium cation with cationic surfactant properties. Further, it is used as an antiseptic and an active pharmaceutical ingredient.

Solubility
Soluble in ethanol, acetone, ethyl acetate, chloroform and water. Slightly soluble in benzene.

Notes
Hygroscopic. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 538-71-6
Molecular Formula C22H40BrNO
Molecular Weight (g/mol) 414.472
MDL Number MFCD00011769
InChI Key OJIYIVCMRYCWSE-UHFFFAOYSA-M
Synonym domiphen bromide, bradasol, bradonit, bradoral, fungitex, oradol, phenododecinium bromide, bradosol bromide, fungitex r
PubChem CID 10866
IUPAC Name dodecyl-dimethyl-(2-phenoxyethyl)azanium;bromide
SMILES CCCCCCCCCCCC[N+](C)(C)CCOC1=CC=CC=C1.[Br-]

Specifications

Melting Point 117°C to 118°C
Quantity 100 g
Beilstein 3922742
Sensitivity Hygroscopic
Merck Index 14,3415
Solubility Information Soluble in ethanol,acetone,ethyl acetate,chloroform and water. Slightly soluble in benzene.
Formula Weight 414.48
Percent Purity 97%
Chemical Name or Material Domiphen bromide
Hazard Category H302
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 208-702-0
RTECSNumber BQ2030000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only