missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dodecylcyclohexane 98.0+%, TCI America™
Spécifications
Spécifications
| Color | Colorless |
| Boiling Point | 126°C |
| Quantity | 500 mL |
| Formula Weight | 252.49 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | Dodecylcyclohexane |
Sécurité et manipulation
| TSCA | Yes |