missing translation for 'onlineSavingsMsg'
Learn More

Dodecylbenzene sulfonic acid sodium salt, 80 to 85%, MP Biomedicals™

Catalog No. ICN15788983
Change view
Click to view available options
Quantity:
250 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15788983 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15788983 Supplier MP Biomedicals Inc Supplier No. 0215788983

Chemical Identifiers

CAS 25155-30-0
Molecular Formula C18H29NaO3S
Molecular Weight (g/mol) 348.48
InChI Key HFQQZARZPUDIFP-UHFFFAOYSA-M
Synonym Sodium dodecylbenzenesulfonate, Abeson nam

Specifications

Color White
Assay Percent Range 80 to 85%
Quantity 250 g
Formula Weight 348.5
Physical Form Solid Flakes
Chemical Name or Material Dodecylbenzene sulfonic acid sodium salt