missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Dodecyl sulfate, sodium salt, 99+%, for molecular biology, DNAse, RNAse and Protease free

CAS: 151-21-3 | C12H25NaO4S | 288.38 g/mol

Supplier:  Thermo Scientific Chemicals 327315000

 View more versions of this product

Catalog No. AC327315000


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

(0.1 M in water), (0.1 M in water) at 0nm, 0.05 max. at 260nm, 0.05 max. at 280nm
White
Powder
500 g
Authentic
C12H25NaO4S
MFCD00036175
Solubility in water: 130g/L (20°C). Other solubilities: partly soluble in alcohol, soluble 75g/L in ethanol (20°C)
[Na+].CCCCCCCCCCCCOS([O-])(=O)=O
288.38
CHEBI:8984
≥99%
Dodecyl sulfate, sodium salt
151-21-3 , 7757-82-6 , 7647-14-5
206.0°C
>150°C
99+%
CH3(CH2)11OSO3Na
Dodecyl Sodium Sulfate, SDS, Sodium lauryl sulfate
DBMJMQXJHONAFJ-UHFFFAOYSA-M
sodium dodecyl sulfate
3423265
288.38
Molecular Biology
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only