missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Dodecyl sulfate, sodium salt, 99+%, for molecular biology, DNAse, RNAse and Protease free

Catalog No. p-124452
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC327311000 100 g
AC327315000 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC327311000 Supplier Thermo Scientific Chemicals Supplier No. 327311000
Only null left

CAS: 151-21-3 | C12H25NaO4S | 288.38 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Name or Material Dodecyl sulfate, sodium salt
Absorbance (0.1 M in water), (0.1 M in water) at 0nm, 0.05 max. at 260nm, 0.05 max. at 280nm
Color White
CAS 151-21-3, 7757-82-6, 7647-14-5
Physical Form Powder
Melting Point 206.0°C
Quantity 100 g
Flash Point >150°C
Infrared Spectrum Authentic
Assay Percent Range 99+%
Molecular Formula C12H25NaO4S
Linear Formula CH3(CH2)11OSO3Na
MDL Number MFCD00036175
Synonym Dodecyl Sodium Sulfate, SDS, Sodium lauryl sulfate
Solubility Information Solubility in water: 130g/L (20°C). Other solubilities: partly soluble in alcohol, soluble 75g/L in ethanol (20°C)
InChI Key DBMJMQXJHONAFJ-UHFFFAOYSA-M
SMILES [Na+].CCCCCCCCCCCCOS([O-])(=O)=O
IUPAC Name sodium;dodecyl sulfate
Molecular Weight (g/mol) 288.38
PubChem CID 3423265
ChEBI CHEBI:8984
Formula Weight 288.38
Percent Purity ≥99%
Grade Molecular Biology
Show More Show Less

RUO – Research Use Only