missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Dodecyl sulfate, sodium salt, 99%, for biochemistry

Catalog No. AC230420100
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
500 g
2.5 kg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC230420100 10 g
AC230420250 25 g
AC230421000 100 g
AC230425000 500 g
AC230420025 2.5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. AC230420100 Supplier Thermo Scientific Chemicals Supplier No. 230420100
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 151-21-3 | C12H25NaO4S, C12H25NaO4S | 288.38 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Denatures and solubilizes proteins for electrophoresis.Sodium n-dodecyl sulfate is an anionic detergent used to function as a protein denaturant. It is mainly used in detergents for laundry with many cleaning applications. It finds application in the determination of protein molecular weight by polyacrylamide gel electrophoresis and in the analysis of hemoglobin. In food products, it is used as an emulsifying agent and a whipping aid. It is widely found in toothpastes, shampoos, shaving creams and bubble bath formulations due to its thickening effect and ability to create a lather.

Solubility
Slightly soluble in ethanol.

Notes
Incompatible with oxidizing agents.

Chemical Identifiers

CAS 151-21-3
Molecular Formula C12H25NaO4S

Specifications

Color White to Beige
Quantity 10 g
Formula Weight 288.38
MDL Number MFCD00036175
Melting Point 206.0°C
Packaging Type Plastic bottle
InChI Key DBMJMQXJHONAFJ-UHFFFAOYSA-M
SMILES [Na+].CCCCCCCCCCCCOS([O-])(=O)=O
IUPAC Name sodium dodecyl sulfate
Molecular Weight (g/mol) 288.38
Purity 99%, Biochemical
Solubility Solubility in water: 130g/L (20°C). Other solubilities: partly soluble in alcohol, soluble 75g/L in ethanol (20°C)
Synonym Sodium lauryl sulfate
Form Crystalline Powder or Crystals
Assay 99%
Chemical Name or Material Dodecyl sulfate, sodium salt
Show More Show Less

RUO – Research Use Only