missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Dodecyl sulfate, sodium salt, 99%, for biochemistry

CAS: 151-21-3 | C12H25NaO4S, C12H25NaO4S | 288.38 g/mol

Supplier:  Thermo Scientific Chemicals 230420025

Quantity 2.5 kg

 View more versions of this product

Catalog No. AC230420025


Only null left

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Denatures and solubilizes proteins for electrophoresis.Sodium n-dodecyl sulfate is an anionic detergent used to function as a protein denaturant. It is mainly used in detergents for laundry with many cleaning applications. It finds application in the determination of protein molecular weight by polyacrylamide gel electrophoresis and in the analysis of hemoglobin. In food products, it is used as an emulsifying agent and a whipping aid. It is widely found in toothpastes, shampoos, shaving creams and bubble bath formulations due to its thickening effect and ability to create a lather.

Solubility
Slightly soluble in ethanol.

Notes
Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

151-21-3
C12H25NaO4S

Specifications

99%
151-21-3
2.5 kg
C12H25NaO4S
206.0°C
DBMJMQXJHONAFJ-UHFFFAOYSA-M
sodium dodecyl sulfate
99%, Biochemical
Sodium lauryl sulfate
Dodecyl sulfate, sodium salt
White to Beige
288.38
MFCD00036175
Plastic bottle
[Na+].CCCCCCCCCCCCOS([O-])(=O)=O
288.38
Solubility in water: 130g/L (20°C). Other solubilities: partly soluble in alcohol, soluble 75g/L in ethanol (20°C)
Crystalline Powder or Crystals
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions