missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dodecyl Alcohol, ACS, 98%, Spectrum™ Chemical
$272.61 - $10,627.11
Chemical Identifiers
| CAS | 112-53-8 |
|---|---|
| Molecular Formula | C12H26O |
| Molecular Weight (g/mol) | 186.34 |
| InChI Key | LQZZUXJYWNFBMV-UHFFFAOYSA-N |
| IUPAC Name | dodecan-1-ol |
| SMILES | CCCCCCCCCCCCO |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
18610310
|
N/A | 100 g | Amber Glass Bottle |
Each for $272.61
|
|
||||
|
18610312
|
N/A | 500 g | Amber Glass Bottle |
Each for $529.29
|
|
||||
|
18610311
|
N/A | 20 L | Amber Glass Bottle |
Each for $4,171.39
|
|
||||
|
18610313
|
N/A | 200 L | Amber Glass Bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
18610309
|
N/A | 3 kg | Amber Glass Bottle |
Each for $1,306.86
|
|
||||
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.