missing translation for 'onlineSavingsMsg'
Learn More

Dodecyl Alcohol, ACS, 98%, Spectrum™ Chemical

Catalog No. 18610310
Change view
Click to view available options
Quantity:
100 g
500 g
3 kg
20 L
200 L
Packaging:
Amber Glass Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18610310 100 g Amber Glass Bottle
18610312 500 g Amber Glass Bottle
18610311 20 L Amber Glass Bottle
18610313 200 L Amber Glass Bottle
18610309 3 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18610310 Supplier Spectrum Chemical Mfg Cor Supplier No. D1067100GM
Get Up to $500 in Chemicals with Purchase
View all promotions

Chemical Identifiers

CAS 112-53-8
Molecular Formula C12H26O
Molecular Weight (g/mol) 186.34
InChI Key LQZZUXJYWNFBMV-UHFFFAOYSA-N
IUPAC Name dodecan-1-ol
SMILES CCCCCCCCCCCCO

Specifications

CAS Max % 1
Boiling Point 254°C to 261°C
Packaging Amber Glass Bottle
Quantity 100 g
Percent Purity 0.98
Grade ACS
Chemical Name or Material Dodecyl Alcohol

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.