missing translation for 'onlineSavingsMsg'
Learn More

Dodecyl Alcohol, ACS, 98%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor D1067500GM

Quantity 500 g
Packaging Amber Glass Bottle

 View more versions of this product

Catalog No. 18610312


Only null left

Description

Description

Specifications

Chemical Identifiers

112-53-8
186.34
dodecan-1-ol
C12H26O
LQZZUXJYWNFBMV-UHFFFAOYSA-N
CCCCCCCCCCCCO

Specifications

Dodecyl Alcohol
1
Amber Glass Bottle
500 g
CCCCCCCCCCCCO
186.34
ACS
112-53-8
254°C to 261°C
C12H26O
LQZZUXJYWNFBMV-UHFFFAOYSA-N
dodecan-1-ol
0.98
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions