accessibility menu, dialog, popup

UserName

H3COH

CAS RN 112-53-8

IUPAC Name: dodecan-1-ol
Synonyms: 1-dodecanol epal 12 lorol 11 epa pesticide chemical code 001509 fema number 2617 hainol 12SS n dodecyl alcohol duodecyl alcohol alcohol C-12 conol 20P
Molecular Weight (g/mol): 186.34
Molecular Formula: C12H26O
InChi Key: LQZZUXJYWNFBMV-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCO
Boiling Point 
  • (5)
  • (3)
  • (3)
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (2)
  • (5)
  • (6)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (3)
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (11)
  • (5)
  • (1)
  • (5)
  • (2)
  • (6)
  • (5)
  • (3)
  • (3)
  • (1)
  • (2)
  • (1)
  • (2)
  • (5)
  • (6)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (3)
  • (5)