missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dodecamethylcyclohexasiloxane 97.0+%, TCI America™
Specifications
Specifications
| Color | Colorless |
| Boiling Point | 245°C |
| Quantity | 1 mL |
| Formula Weight | 444.92 |
| Percent Purity | ≥97.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | Dodecamethylcyclohexasiloxane |
Safety and Handling
| RTECSNumber | GW2328550 |
| TSCA | Yes |