missing translation for 'onlineSavingsMsg'
Learn More

Doctors Solution, Sodium Plumbite, Certified, 3.2 to 3.5N, LabChem™

Supplier:  LabChem LC136904

 View more versions of this product

Catalog No. LC136904


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Doctors Solution
Sodium Plumbite
1310-73-2 , 1317-36-8
PbO
1.1g/mL
Soluble in water
Certified
Poly Bottle
Traceable to NIST
Fume; Lead oxide; Thermal decomposition generates: Corrosive vapors
4 L
Colorless
86.4,11.3,2.3
3.1 to 3.3N at pH 11.0
3.2 to 3.5N
water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide
223.19
Passes Test
1.1g/mL
1.8cSt
7.0
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions