missing translation for 'onlineSavingsMsg'
Learn More

Doctors Solution, Sodium Plumbite, Certified, 3.2 to 3.5N, LabChem™

Catalog No. p-5388384
Change view
Click to view available options
Quantity:
1 L
4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC136902 1 L
LC136904 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. LC136902 Supplier LabChem Supplier No. LC136902

Specifications

Chemical Name or Material Doctors Solution
Color Colorless
Name Note Sodium Plumbite
CAS Max % 86.4, 11.3, 2.3
CAS 1310-73-2, 1317-36-8
Assay Percent Notes 3.1 to 3.3N at pH 11.0
Linear Formula PbO
Concentration 3.2 to 3.5N
Density 1.1g/mL
Synonym water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide
Solubility Information Soluble in water
Formula Weight 223.19
Grade Certified
Identification Passes Test
Packaging Poly Bottle
Specific Gravity 1.1g/mL
Traceability to NIST Traceable to NIST
Viscosity 1.8cSt
Decomposition Information Fume; Lead oxide; Thermal decomposition generates: Corrosive vapors
pH 7.0
Quantity 1 L
Physical Form Liquid
Show More Show Less