missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ DNA Polymerase I (10 U/μL)

Catalog No. FEREP0042
Change view
Click to view available options
Quantity:
2,500 units
500 units
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEREP0042 2,500 units
FEREP0041 500 units
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. FEREP0042 Supplier Thermo Scientific™ Supplier No. EP0042

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

DNA Polymerase I is a template-dependent DNA polymerase which catalyzes 5'→3' synthesis of DNA.

DNA Polymerase I, a template-dependent DNA polymerase, catalyzes 5'→3' synthesis of DNA. The enzyme also exhibits 3'→5' exonuclease (proofreading) activity, 5'→3' exonuclease activity, and ribonuclease H activity.

Highlights

  • Incorporates modified nucleotides(e.g. biotin-, digoxigenin-, aminoallyl-, fluorescently-labeled nucleotides)
  • Active in multiple buffers, including restriction enzyme, PCR, and RT buffers

Applications

  • DNA labeling by nick-translation in conjunction with DNase
  • Second-strand synthesis of cDNA in conjunction with RNase

Specifications

Concentration 10 U/μL
Description DNA Polymerase I
Polymerase DNA Polymerase I
Quantity 2,500 units
What are the differences between Thermo Scientific DNA Polymerase I, Klenow Fragment, and Klenow Fragment (exo-)?

Thermo Scientific DNA Polymerase I possesses 5′?3′ DNA synthesis activity, 3′?5′ exonuclease (proofreading) activity, and 5′?3′ exonuclease activity. Klenow Fragment is the large fragment of DNA polymerase I. It exhibits 5′?3′ polymerase activity and 3′?5′ exonuclease (proofreading) activity, but lacks 5′?3′ exonuclease activity. Klenow Fragment, exo-, is also the large fragment of DNA polymerase I. It exhibits 5′?3′ polymerase activity, but lacks the 3′?5′ and 5′?3′ exonuclease activities.

For Research Use Only. Not for use in diagnostic procedures.