missing translation for 'onlineSavingsMsg'
Learn More

Promega DNA Polymerase I

Numéro de catalogue. PRM2055
Encompass
Change view
Click to view available options
Quantity:
2500 U
500 U
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
PRM2055 2500 U
PRM2051 500 U
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. PRM2055 Fournisseur Promega Code fournisseur M2055
Il en reste null

DNA Polymerase I catalyzes the template-directed polymerization of nucleotides into duplex DNA in a 5'→3' direction. Recombinant.

  • Catalyzes the Template-Directed Polymerization of Nucleotides into Duplex DNA in a 5′→3′ Direction
  • Can be used in a variety of molecular applications
  • Is inactivated by heating at 68°C for 10 minutes
  • Supplied at a concentration of 5-10u/μL

Spécifications

Concentration 5 to 10 U/μL
Color Colorless
Components 10X Reaction Buffer: 500mM Tris HCl (pH 7.2 at 25°C), 100mM MgSO4, DTT 1mM
Purity >90%
pH 7.5
For Use With (Application) Labeling of DNA to high specific radioactivity by nick translation, second-strand cDNA synthesis
Storage Buffer 50mM Tris HCl (pH 7.5 at 25°C), 1mM DTT, 0.1mM EDTA, 50% Glycerol
Content And Storage -30°C to -10°C
Source Recombinant E. coli strain
Quantity 2500 U
Product Type DNA Polymerase I
Form Liquid
Afficher plus de résultats Afficher moins de résultats