missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human DNA-PK Native Protein
Description
For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Native human protein purified from MOLT-4 cells. No special measures were taken to activate this kinase. Protein Form: Full Length.
DNA-PK is a heterotrimeric, DNA-dependent protein kinase that initiates signaling cascades in response to DNA damage. DNA-PK is an atypical kinase, closely related to the PI3 kinases, and is often potently inhibited by small molecules designed to target PI3 kinases. DNA-PK encodes a component of the mediator complex, a transcriptional coactivator complex thought to be required for the expression of almost all genes. The mediator complex is recruited by transcriptional activators or nuclear receptors to induce gene expression, possibly by interacting with RNA polymerase II and promoting the formation of a transcriptional pre-initiation complex. Multiple transcript variants encoding different isoforms have been found for DNA-PK.
Spécifications
Spécifications
| Numéro d'enregistrement | P78527 |
| Concentration | Conc. Not Determined |
| À utiliser avec (application) | Kinase Assay |
| Formule | 25mM HEPES with 10mM MgCl2, 50mM KCl, 1mM DTT, 0.2mM EDTA, 10% glycerol and no preservative; pH 7.5 |
| ID du gène (Entrez) | 5591 |
| Poids moléculaire (g/mol) | 600 kDa |
| Nom | Human DNA-PK |
| Plage de pH | 7.5 |
| Quantité | 100 μg |
| Conditions de stockage | -80°C, Avoid Freeze/Thaw Cycles |
| Afficher plus de résultats |