missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ DNA Molecular Weight Marker, Wide Range Ladder (100-12,000 bp)

Numéro de catalogue. AAJ67051AMI
Modifier l'affichage
Click to view available options
Quantité:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité
AAJ67051AMI 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. AAJ67051AMI Fournisseur Thermo Scientific™ Code fournisseur J67051AMI

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

 

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Spécifications

Format Liquid
Quantité 0.5 mL
Conditions de stockage 2° to 8°C

RUO – Research Use Only