missing translation for 'onlineSavingsMsg'
Learn More

DMF, Dimethylformamide, Sequencing, Fisher BioReagents

Catalog No. BP11604
Change view
Click to view available options
Quantity:
4 L
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
BP11604 4 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BP11604 Supplier Fisher BioReagents Supplier No. BP11604
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

  • DMF for sequencing and peptide synthesis
  • Useful for protein chemistry due to low amine content
  • DMF is a commonly used solvent in solid phase peptide synthesis
  • Intended for research use only

Chemical Identifiers

CAS 68-12-2
Molecular Formula C3H7NO
Molecular Weight (g/mol) 73.10
MDL Number MFCD00003284
InChI Key ZMXDDKWLCZADIW-UHFFFAOYSA-N
Synonym dimethylformamide, dimethyl formamide, n,n-dimethylmethanamide, n-formyldimethylamine, formamide, n,n-dimethyl, dmf, dimethylformamid, dimetilformamide, dwumetyloformamid, dmfa
PubChem CID 6228
ChEBI CHEBI:17741
IUPAC Name N,N-dimethylformamide
SMILES CN(C)C=O

Specifications

pH 6 to 8
Melting Point -61°C
Absorbance Pass Test, 1 max. at 270nm, 0.3 max. at 275nm, 0.1 max. at 295nm, 0.05 max. at 310nm, 0.01 max. (340nm to 400nm) at 340nm
Color Colorless
Decomposition Information >350°C
Density 0.945g/cm³
Quantity 4 L
Identification Pass Test
Packaging Glass Bottle
Physical Form Liquid
Residue after Evaporation 0.005% max.
Assay Percent Range ≥99.5 %
Titratable Acid 0.0005mEq/g max.
Titratable Base 0.003mEq/g max.
Vapor Pressure 4.9mbar at 20°C
Linear Formula HCONCH3CH3
Viscosity 0.8 mPa/s at 20°C
UN Number 2265
Water 0.03% max.
Merck Index 15, 3267
Boiling Point 153°C
Formula Weight 73.09
Percent Purity ≥99.5%
Grade Sequencing
Chemical Name or Material N, N-Dimethylformamide
Show More Show Less
Safety handling Solvent
Hazard Statement WARNING!
Precautionary Statement Emergency Overview
Flammable liquid and vapor. May cause methemoglobinemia. Irritating to eyes and skin. Lachrymator (substance which increases the flow of tears). May cause central nervous system effects. May cause adverse liver effects. May cause adverse kidney effects. May cause harm to the unborn child. Use personal protective equipment. Keep away from open flames
DOTInformation DOT Class 3, : Flammable Liquid
Recommended Storage RT