missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ DMEM for SILAC

Numéro de catalogue. p-7083440
Change view
Click to view available options
Quantity:
500 mL
6 x 500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
PIA33822 6 x 500 mL
PI88364 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. PIA33822 Fournisseur Thermo Scientific™ Code fournisseur A33822
Il en reste null

Analyze protein expression by mass spectrometry using these media or additives for stable isotope labeling with amino acids in cell culture (SILAC).

Thermo Scientific™ DMEM for SILAC is optimized for use with stable isotope labeling with amino acids in cell culture (SILAC) to analyze protein expression by mass spectrometry (MS).

Features of DMEM for SILAC:

  • Flexible—liquid media deficient in both L-lysine and L-arginine, allowing for more complete proteome coverage through multiple isotopic amino acid labeling
  • High quality—medium is sterile, endotoxin-free, and cell culture-compatible

SILAC is a powerful method to identify and quantify relative differential changes in complex protein samples. This approach entails the in vivo metabolic incorporation of 'heavy' 13C- or 15N-labeled amino acids into proteins followed by mass spectrometry (MS) for the accelerated and comprehensive identification, characterization, and quantitation of proteins.

This SILAC medium is used together with heavy and light amino acids for the differential isotopic labeling of cells for SILAC analysis. SILAC media supplemented with heavy amino acids are capable of nearly 100% label incorporation into proteins of living cells.

Related products

  • NeuCode™ Lysine -080, 50 mg (A33613)
  • Fetal Bovine Serum, dialyzed, US origin (26400036)
  • RPMI 1640 Media for SILAC (88365)

Spécifications

For Use With (Equipment) Mass Spectrometer
Product Type Media
Content And Storage Store at 2°C to 8°C.
Label or Dye Mass Isotope of Amino Acid
Shipping Condition Room Temperature
Starting Material Cell Lysate
Final Product Type Liquid Medium
Quantity 6 x 500 mL
Form Liquid

For Research Use Only. Not for use in diagnostic procedures.