missing translation for 'onlineSavingsMsg'
Learn More

DL-Xylose 98.0+%, TCI America™

Numéro de catalogue. X00201G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
X00201G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. X00201G Fournisseur TCI America Code fournisseur X00201G
Il en reste null

Identifiants chimiques

CAS 25990-60-7
Molecular Formula C5H10O5
Molecular Weight (g/mol) 150.13
MDL Number MFCD00198055
InChI Key PYMYPHUHKUWMLA-VPENINKCSA-N
PubChem CID 644160
ChEBI CHEBI:15936
IUPAC Name (2R,3S,4R)-2,3,4,5-tetrahydroxypentanal
SMILES C(C(C(C(C=O)O)O)O)O

Spécifications

Melting Point 136°C
Color White
Quantity 1 g
Formula Weight 150.13
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material DL-Xylose
EINECSNumber (9)-0318
TSCA No