missing translation for 'onlineSavingsMsg'
Learn More

DL-Valine, Thermo Scientific™

Chemical Identifiers

CAS 516-06-3
Molecular Formula C5H11NO2
Molecular Weight (g/mol) 117.15
MDL Number MFCD00004267
InChI Key KZSNJWFQEVHDMF-UHFFFAOYNA-N
Synonym dl-valine, h-dl-val-oh, valin, valine, dl, dl-2-amino-3-methylbutanoic acid, 2-amino-3-methylbutanoic acid, dl, valina, dl-.alpha.-aminoisovaleric acid, dl-2-amino-3-methylbutyric acid, 2-aminoisovaleric acid
PubChem CID 1182
ChEBI CHEBI:27266
IUPAC Name 2-amino-3-methylbutanoic acid
SMILES CC(C)C(N)C(O)=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
AC452832500
View Documents Promotion Details
N/A 250g Plastic bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC452830010
View Documents Promotion Details
N/A 1kg Plastic bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

516-06-3
117.15
KZSNJWFQEVHDMF-UHFFFAOYNA-N
1182
2-amino-3-methylbutanoic acid
C5H11NO2
MFCD00004267
dl-valine, h-dl-val-oh, valin, valine, dl, dl-2-amino-3-methylbutanoic acid, 2-amino-3-methylbutanoic acid, dl, valina, dl-.alpha.-aminoisovaleric acid, dl-2-amino-3-methylbutyric acid, 2-aminoisovaleric acid
CHEBI:27266
CC(C)C(N)C(O)=O

Specifications

516-06-3
MFCD00004267
KZSNJWFQEVHDMF-UHFFFAOYNA-N
2-amino-3-methylbutanoic acid
1182
117.15
DL-Valine
C5H11NO2
dl-valine, h-dl-val-oh, valin, valine, dl, dl-2-amino-3-methylbutanoic acid, 2-amino-3-methylbutanoic acid, dl, valina, dl-.alpha.-aminoisovaleric acid, dl-2-amino-3-methylbutyric acid, 2-aminoisovaleric acid
CC(C)C(N)C(O)=O
117.15
CHEBI:27266
Extra Pure
Videos
SDS
Documents

Documents

RUO – Research Use Only