missing translation for 'onlineSavingsMsg'
Learn More

DL-Valine Methyl Ester Hydrochloride 98.0+%, TCI America™

Catalog No. V01255G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
V01255G 5 g
V012525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. V01255G Supplier TCI America Supplier No. V01255G

Chemical Identifiers

CAS 5619-05-6
Molecular Formula C6H14ClNO2
Molecular Weight (g/mol) 167.63
MDL Number MFCD00066118
InChI Key KUGLDBMQKZTXPW-UHFFFAOYSA-N
Synonym dl-valine methyl ester hydrochloride, h-dl-val-ome.hcl, methyl 2-amino-3-methylbutanoate hydrochloride, h-d-val-ome.hcl, methyl dl-valinate hydrochloride, d,l-valine methyl ester hydrochloride, h-d-val-ome*hcl, h-val-ome??hcl, acmc-209fna, acmc-209ncl
PubChem CID 12498813
IUPAC Name methyl 2-amino-3-methylbutanoate hydrochloride
SMILES Cl.COC(=O)C(N)C(C)C

Specifications

Melting Point 105°C
Color White
Quantity 5 g
Formula Weight 167.63
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material DL-Valine Methyl Ester Hydrochloride